missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB39B Protein
A cDNA sequence encoding the RAB39B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13240-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
RAB39B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEAIWLY QFRLIVIGDS TVGKSCLIRR FTEGRFAQVS DPTVGVDFFS RLVEIEPGKR IKLQIWDTAG QERFRSITRA YYRNSVGGLL LFDITNRRSF QNVHEWLEET KVHVQPYQIV FVLVGHKCDL DTQRQVTRHE AEKLAAAYGM KYIETSARDA INVEKAFTDL TRDIYELVKR GEITIQEGWE GVKSGFVPNV VHSSEEVVKS ERRCLC | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAB39B protein solution (0.25 mg/ml) contains in Phosphate buffered saline (pH 7.4), 30% glycerol and 1mM DTT. |