missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAF1 Protein
A DNA sequence encoding the Homo sapiens (Human) RAF1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6581-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002871.1 | |
RAF1 Protein | |
Research Use Only | |
RAF1 | |
Recombinant Protein | |
E. coli | |
MEHIQGAWKT ISNGFGFKDA VFDGSSCISP TIVQQFGYQR RASDDGKLTD PSKTSNTIRV FLPNKQRTVV NVRNGMSLHD CLMKALKVRG LQPECCAVFR LLHEHKGKKA RLDWNTDAAS LIGEELQVDF LDHVPLTTHN FARKTFLKLA FCDICQKFLL NGFRCQTCGY KFHEHCSTKV PTMCVDWSNI RQLLLFPNST IGDSGVPALP SLTMRRMRES VSRMPVSSQH RYSTPHAFTF NTSSPSSEGS LSQRQRSTST PNVHMVSTTL PVDSRMIEDA IRSHSESASP SALSSSPNNL SPTGWSQPKT PVPAQRERAP VSGTQEKNKI RPRGQRDSSY YWEIEASEVM LSTRIGSGSF GTVYKGKWHG DVAVKILKVV DPTPEQFQAF RNEVAVLRKT RHVNILLFMG YMTKDNLAIV TQWCEGSSLY KHLHVQETKF QMFQLIDIAR QTAQGMDYLH AKNIIHRDMK SNNIFLHEGL TVKIGDFGLA TVKSRWSGSQ QVEQPTGSVL WMAPEVIRMQ DNNPFSFQSD VYSYGIVLYE LMTGELPYSH INNRDQIIFM VGRGYASPDL SKLYKNCPKA MKRLVADCVK KVKEERPLFP QILSSIELLQ HSLPKINRSA SEPSLHRAAH TEDINACTLT TSPRLPVF | |
Greater than 90% as determined by SDS-PAGE. |
5894 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |