missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-18 Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-18, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8149-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001243339.1 | |
Ras-related protein Rab-18 Protein | |
Research Use Only | |
RAB18 | |
Recombinant Protein | |
E. coli | |
MDEDVLTTLK ILIIGESGVG KSSLLLRFTD DTFDPELAAT IGVDFKVKTI SVDGNKAKLA IWDTAGQERF RTLTPSYYRG AQGVILVYDV TRRDTFVKLD NWLNELETYC TRNDIVNMLV GNKIDKENRE VDRNEGLKFA RKHSMLFIEA SAKTCDGVQC AFEELVEKII QTPGLWESEN QNKGVKLSHR EEGQGGGACG GYC | |
Greater than 80% as determined by SDS-PAGE. |
22931 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |