missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-1A Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-1A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8316-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_004152.1 | |
Ras-related protein Rab-1A Protein | |
Research Use Only | |
RAB1A | |
Recombinant Protein | |
E. coli | |
SSMNPEYDYL FKLLLIGDSG VGKSCLLLRF ADDTYTESYI STIGVDFKIR TIELDGKTIK LQIWDTAGQE RFRTITSSYY RGAHGIIVVY DVTDQESFNN VKQWLQEIDR YASENVNKLL VGNKCDLTTK KVVDYTTAKE FADSLGIPFL ETSAKNATNV EQSFMTMAAE IKKRMGPGAT AGGAEKSNVK IQSTPVKQSG GGCC | |
Greater than 90% as determined by SDS-PAGE. |
5861 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |