missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Ral-B Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Ral-B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6582-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002872.1 | |
Ras-related protein Ral-B Protein | |
Research Use Only | |
RALB | |
Recombinant Protein | |
E. coli | |
MAANKSKGQS SLALHKVIMV GSGGVGKSAL TLQFMYDEFV EDYEPTKADS YRKKVVLDGE EVQIDILDTA GQEDYAAIRD NYFRSGEGFL LVFSITEHES FTATAEFREQ ILRVKAEEDK IPLLVVGNKS DLEERRQVPV EEARSKAEEW GVQYVETSAK TRANVDKVFF DLMREIRTKK MSENKDKNGK KSSKNKKSFK ERC | |
Greater than 80% as determined by SDS-PAGE. |
5899 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |