missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RCAS1 / EBAG9 Protein
A DNA sequence encoding the Homo sapiens (Human) RCAS1 / EBAG9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5959-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001265867.1 | |
RCAS1 / EBAG9 Protein | |
Research Use Only | |
EBAG9 | |
Recombinant Protein | |
E. coli | |
RSGRGRKLSG DQITLPTTVD YSSVPKQTDV EEWTSWDEDA PTSVKIEGGN GNVATQQNSL EQLEPDYFKD MTPTIRKTQK IVIKKREPLN FGIPDGSTGF SSRLAATQDL PFIHQSSELG DLDTWQENTN AWEEEEDAAW QAEEVLRQQK LADREKRAAE QQRKKMEKEA QRLMKKEQNK IGVKLS | |
Greater than 90% as determined by SDS-PAGE. |
9166 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |