missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RGS5 / Regulator of G-protein signaling 5 Protein
A DNA sequence encoding the Homo sapiens (Human) RGS5 / Regulator of G-protein signaling 5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6605-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001182232.1 | |
RGS5 / Regulator of G-protein signaling 5 Protein | |
Research Use Only | |
RGS5 | |
Recombinant Protein | |
E. coli | |
MCKGLAALPH SCLERAKEIK IKLGILLQKP DSVGDLVIPY NEKPEKPAKT QKTSLDEALQ WRDSLDKLLQ NNYGLASFKS FLKSEFSEEN LEFWIACEDY KKIKSPAKMA EKAKQIYEEF IQTEAPKEVN IDHFTKDITM KNLVEPSLSS FDMAQKRIHA LMEKDSLPRF VRSEFYQELI K | |
Greater than 80% as determined by SDS-PAGE. |
8490 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |