missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rho guanine nucleotide exchange factor 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Rho guanine nucleotide exchange factor 7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8369-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001106983.1 | |
Rho guanine nucleotide exchange factor 7 Protein | |
Research Use Only | |
ARHGEF7 | |
Recombinant Protein | |
E. coli | |
MNSAEQTVTW LITLGVLESP KKTISDPEGF LQASLKDGVV LCRLLERLLP GTIEKVYPEP RSESECLSNI REFLRGCGAS LRLELLFPPS QPPQHLVTTI LLSASTFDAN DLYQGQNFNK VLSSLVTLNK VTADIGLGSD SVCARPSSHR IKSFDSLGSQ SLHTRTSKLF QGQYRSLDMT DNSNNQLVVR AKFNFQQTNE DELSFSKGDV IHVTRVEEGG WWEGTLNGRT GWFPSNYVRE VKASEKPVSP | |
Greater than 90% as determined by SDS-PAGE. |
8874 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |