missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RhoA Protein
A DNA sequence encoding the Homo sapiens (Human) RhoA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8336-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001300870.1 | |
RhoA Protein | |
Research Use Only | |
RHOA | |
Recombinant Protein | |
E. coli | |
MAAIRKKLVI VGDGACGKTC LLIVFSKDQF PEVYVPTVFE NYVADIEVDG KQVELALWDT AGQEDYDRLR PLSYPDTDVI LMCFSIDSPD SLENIPEKWT PEVKHFCPNV PIILVGNKKD LRNDEHTRRE LAKMKQEPVK PEEGRDMANR IGAFGYMECS AKTKDGVREV FEMATRAALQ ARRGKKKSGC L | |
Greater than 90% as determined by SDS-PAGE. |
387 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |