missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rhombotin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Rhombotin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8396-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001257357.1 | |
Rhombotin-1 Protein | |
Research Use Only | |
LMO1 | |
Recombinant Protein | |
E. coli | |
DKEDGVPMLS VQPKGKQKGC AGCNRKIKDR YLLKALDKYW HEDCLKCACC DCRLGEVGST LYTKANLILC RRDYLRLFGT TGNCAACSKL IPAFEMVMRA RDNVYHLDCF ACQLCNQRFC VGDKFFLKNN MILCQMDYEE GQLNGTFESQ VQ | |
Greater than 90% as determined by SDS-PAGE. |
4004 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |