missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A8 Protein
A cDNA sequence encoding the S100A8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13399-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
S100A8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS MLTELEKALN SIIDVYHKYS LIKGNFHAVY RDDLKKLLET ECPQYIRKKG ADVWFKELDI NTDGAVNFQEMLTELEKALN SIIDVYHKYS LIKGNFHAVY RDDLKKLLET ECPQYIRKKG ADVWFKELDI NTDGAVNFQE FLILVIKMGV AAHKKSHEES HKE |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
S100A8 was filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM Tris and 100mM NaCl, pH 7.5. |