missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SDSL Protein
A cDNA sequence encoding the SDSL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13444-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
113675 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SDSL | |
Human | |
His | |
The SDSL solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 0.1M NaCl and 10% glycerol. |
SDSL Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMDGPVA EHAKQEPFHV VTPLLESWAL SQVAGMPVFL KCENVQPSGS FKIRGIGHFC QEMAKKGCRH LVCSSGGNAG IAAAYAARKL GIPATIVLPE STSLQVVQRL QGEGAEVQLT GKVWDEANLR AQELAKRDGW ENVPPFDHPL IWKGHASLVQ ELKAVLRTPP GALVLAVGGG GLLAGVVAGL LEVGWQHVPI IAMETHGAHC FNAAITAGKL VTLPDITSVA KSLGAKTVAA RALECMQVCK IHSEVVEDTE AVSAVQQLLD DERMLVEPAC GAALAAIYSG LLRRLQAEGC LPPSLTSVVV IVCGGNNINS RELQALKTHL GQV |