missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Septin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Septin-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7552-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001008491.1 | |
Septin-2 Protein | |
Research Use Only | |
42980 | |
Recombinant Protein | |
E. coli | |
MSKQQPTQFI NPETPGYVGF ANLPNQVHRK SVKKGFEFTL MVVGESGLGK STLINSLFLT DLYPERVIPG AAEKIERTVQ IEASTVEIEE RGVKLRLTVV DTPGYGDAIN CRDCFKTIIS YIDEQFERYL HDESGLNRRH IIDNRVHCCF YFISPFGHGL KPLDVAFMKA IHNKVNIVPV IAKADTLTLK ERERLKKRIL DEIEEHNIKI YHLPDAESDE DEDFKEQTRL LKASIPFSVV GSNQLIEAKG KKVRGRLYPW GVVEVENPEH NDFLKLRTML ITHMQDLQEV TQDLHYENFR SERLKRGGRK VENEDMNKDQ ILLEKEAELR RMQEMIARMQ AQMQMQMQGG DGDGGALGHH V | |
Greater than 90% as determined by SDS-PAGE. |
4735 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |