missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine protease HTRA1 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine protease HTRA1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7864-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002766.1 | |
Serine protease HTRA1 Protein | |
Research Use Only | |
HTRA1 | |
Recombinant Protein | |
E. coli | |
QLSRAGRSAP LAAGCPDRCE PARCPPQPEH CEGGRARDAC GCCEVCGAPE GAACGLQEGP CGEGLQCVVP FGVPASATVR RRAQAGLCVC ASSEPVCGSD ANTYANLCQL RAASRRSERL HRPPVIVLQR GACGQGQEDP NSLRHKYNFI ADVVEKIAPA VVHIELFRKL PFSKREVPVA SGSGFIVSED GLIVTNAHVV TNKHRVKVEL KNGATYEAKI KDVDEKADIA LIKIDHQGKL PVLLLGRSSE LRPGEFVVAI GSPFSLQNTV TTGIVSTTQR GGKELGLRNS DMDYIQTDAI INYGNSGGPL VNLDGEVIGI NTLKVTAGIS FAIPSDKIKK FLTESHDRQA KGKAITKKKY IGIRMMSLTS SKAKELKDRH RDFPDVISGA YIIEVIPDTP AEAGGLKEND VIISINGQSV VSANDVSDVI KRESTLNMVV RRGNEDIMIT VIPEEIDP | |
Greater than 90% as determined by SDS-PAGE. |
5654 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |