missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serum amyloid P component / APCS / SAP Protein
A DNA sequence encoding the Homo sapiens (Human) Serum amyloid P component / APCS / SAP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5659-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001630.1 | |
Serum amyloid P component / APCS / SAP Protein | |
Research Use Only | |
APCS | |
Recombinant Protein | |
E. coli | |
HTDLSGKVFV FPRESVTDHV NLITPLEKPL QNFTLCFRAY SDLSRAYSLF SYNTQGRDNE LLVYKERVGE YSLYIGRHKV TSKVIEKFPA PVHICVSWES SSGIAEFWIN GTPLVKKGLR QGYFVEAQPK IVLGQEQDSY GGKFDRSQSF VGEIGDLYMW DSVLPPENIL SAYQGTPLPA NILDWQALNY EIRGYVIIKP LVWV | |
Greater than 80% as determined by SDS-PAGE. |
325 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |