missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIRT3 Protein
A cDNA sequence encoding the SIRT3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13509-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
SIRT3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSDKGKLSLQ DVAELIRARA CQRVVVMVGA GISTPSGIPD FRSPGSGLYS NLQQYDLPYP EAIFELPFFF HNPKPFFTLA KELYPGNYKP NVTHYFLRLL HDKGLLLRLY TQNIDGLERV SGIPASKLVE AHGTFASATC TVCQRPFPGE DIRADVMADR VPRCPVCTGV VKPDIVFFGE PLPQRFLLHV VDFPMADLLL ILGTSLEVEP FASLTEAVRS SVPRLLINRD LVGPLAWHPR SRDVAQLGDV VHGVESLVEL LGWTEEMRDL VQRETGKLDG PDK | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SIRT3 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 1mM DTT, 0.1M NaCl, 10% glycerol. |