missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNAI2 Protein
A cDNA sequence encoding the SNAI2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13530-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
SNAI2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPRSFLV KKHFNASKKP NYSELDTHTV IISPYLYESY SMPVIPQPEI LSSGAYSPIT VWTTAAPFHA QLPNGLSPLS GYSSSLGRVS PPPPSDTSSK DHSGSESPIS DEEERLQSKL SDPHAIEAEK FQCNLCNKTY STFSGLAKHK QLHCDAQSRK SFSCKYCDKE YVSLGALKMH IRTHTLPCVC KICGKAFSRP WLLQGHIRTH TGEKPFSCPH CNRAFADRSN LRAHLQTHSD VKKYQCKNCS KTFSRMSLLH KHEESGCCVA H | |
Greater than 80.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNAI2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl Buffer (pH 8.0) and 10% glycerol. |