missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Stathmin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Stathmin-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8172-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001269982.1 | |
Stathmin-4 Protein | |
Research Use Only | |
STMN4 | |
Recombinant Protein | |
E. coli | |
MTLAAYKEKM KELPLVSLFC SCFLADPLNK SSYKYEGWCG RQCRRKDESQ RKDSADWRER RAQADTVDLN WCVISDMEVI ELNKCTSGQS FEVILKPPSF DGVPEFNASL PRRRDPSLEE IQKKLEAAEE RRKYQEAELL KHLAEKREHE REVIQKAIEE NNNFIKMAKE KLAQKMESNK ENREAHLAAM LERLQEKDKH AEEVRKNKEL KEEASR | |
Greater than 80% as determined by SDS-PAGE. |
81551 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |