Learn More
enQuireBio™ Recombinant Human Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
A DNA sequence encoding the Homo sapiens (Human) Steroid 17-alpha-hydroxylase/17,20 lyase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5905-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000093.1 | |
Steroid 17-alpha-hydroxylase / 17,20 lyase Protein | |
Research Use Only | |
CYP17A1 | |
Recombinant Protein | |
E. coli | |
GAKYPKSLLS LPLVGSLPFL PRHGHMHNNF FKLQKKYGPI YSVRMGTKTT VIVGHHQLAK EVLIKKGKDF SGRPQMATLD IASNNRKGIA FADSGAHWQL HRRLAMATFA LFKDGDQKLE KIICQEISTL CDMLATHNGQ SIDISFPVFV AVTNVISLIC FNTSYKNGDP ELNVIQNYNE GIIDNLSKDS LVDLVPWLKI FPNKTLEKLK SHVKIRNDLL NKILENYKEK FRSDSITNML DTLMQAKMNS DNGNAGPDQD SELLSDNHIL TTIGDIFGAG VETTTSVVKW TLAFLLHNPQ VKKKLYEEID QNVGFSRTPT ISDRNRLLLL EATIREVLRL RPVAPMLIPH KANVDSSIGE FAVDKGTEVI INLWALHHNE KEWHQPDQFM PERFLNPAGT QLISPSVSYL PFGAGPRSCI GEILARQELF LIMAWLLQRF DLEVPDDGQL PSLEGIPKVV FLIDSFKVKI KVRQAWREAQ AEGST | |
Greater than 80% as determined by SDS-PAGE. |
1586 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |