missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STMN3 Protein
A cDNA sequence encoding the STMN3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13622-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
STMN3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDMEVKQLDK RASGQSFEVI LKSPSDLSPE SPMLSSPPKK KDTSLEELQK RLEAAEERRK TQEAQVLKQL AERREHEREV LHKALEENNN FSRQAEEKLN YKMELSKEIR EAHLAALRER LREKELHAAE VRRNKEQREE MSG | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STMN3 Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |