missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STX1A Protein

A cDNA sequence encoding the STX1A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13632-25ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15954209

  • 181.00€ / 25µg

Spedizione stimata: 06-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

6804
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
STX1A
Human
His
The protein solution contains 20mM Tris-HCl pH 7.5, 10% glycerol, and 1mM DTT.
STX1A Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MKDRTQELRT AKDSDDDDDV AVTVDRDRFM DEFFEQVEEI RGFIDKIAEN VEEVKRKHSA ILASPNPDEK TKEELEELMS DIKKTANKVR SKLKSIEQSI EQEEGLNRSS ADLRIRKTQH STLSRKFVEV MSEYNATQSD YRERCKGRIQ RQLEITGRTT TSEELEDMLE SGNPAIFASG IIMDSSISKQ ALSEIETRHS EIIKLENSIR ELHDMFMDMA MLVESQ
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali