missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-cell surface glycoprotein CD1c Protein
A DNA sequence encoding the Homo sapiens (Human) T-cell surface glycoprotein CD1c, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5783-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001756.2 | |
T-cell surface glycoprotein CD1c Protein | |
Research Use Only | |
CD1C | |
Recombinant Protein | |
Yeast | |
NADASQEHVS FHVIQIFSFV NQSWARGQGS GWLDELQTHG WDSESGTIIF LHNWSKGNFS NEELSDLELL FRFYLFGLTR EIQDHASQDY SKYPFEVQVK AGCELHSGKS PEGFFQVAFN GLDLLSFQNT TWVPSPGCGS LAQSVCHLLN HQYEGVTETV YNLIRSTCPR FLLGLLDAGK MYVHRQVRPE AWLSSRPSLG SGQLLLVCHA SGFYPKPVWV TWMRNEQEQL GTKHGDILPN ADGTWYLQVI LEVASEEPAG LSCRVRHSSL GGQDIILYWG HHFSM | |
Greater than 90% as determined by SDS-PAGE. |
911 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |