missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human T-cell surface glycoprotein CD1c Protein

A DNA sequence encoding the Homo sapiens (Human) T-cell surface glycoprotein CD1c, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5783-ec-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15947719

  • 680.00€ / 100µg

Spedizione stimata: 27-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

NP_001756.2
T-cell surface glycoprotein CD1c Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
911
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD1C
Recombinant Protein
E. coli
NADASQEHVS FHVIQIFSFV NQSWARGQGS GWLDELQTHG WDSESGTIIF LHNWSKGNFS NEELSDLELL FRFYLFGLTR EIQDHASQDY SKYPFEVQVK AGCELHSGKS PEGFFQVAFN GLDLLSFQNT TWVPSPGCGS LAQSVCHLLN HQYEGVTETV YNLIRSTCPR FLLGLLDAGK MYVHRQVRPE AWLSSRPSLG SGQLLLVCHA SGFYPKPVWV TWMRNEQEQL GTKHGDILPN ADGTWYLQVI LEVASEEPAG LSCRVRHSSL GGQDIILYWG HHFSM
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali