Learn More
enQuireBio™ Recombinant Human Tetratricopeptide repeat protein 1
A DNA sequence encoding the Homo sapiens (Human) Tetratricopeptide repeat protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8348-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001269429.1 | |
Tetratricopeptide repeat protein 1 | |
Research Use Only | |
TTC1 | |
Recombinant Protein | |
E. coli | |
EKSENCGVPE DLLNGLKVTD TQEAECAGPP VPDPKNQHSQ SKLLRDDEAH LQEDQGEEEC FHDCSASFEE EPGADKVENK SNEDVNSSEL DEEYLIELEK NMSDEEKQKR REESTRLKEE GNEQFKKGDY IEAESSYSRA LEMCPSCFQK ERSILFSNRA AARMKQDKKE MAINDCSKAI QLNPSYIRAI LRRAELYEKT DKLDEALEDY KSILEKDPSI HQAREACMRL PKQIEERNER LKEEMLGKLK DLGNLVLRPF GLSTENFQIK QDSSTGSYSI NFVQNPNNNR | |
Greater than 90% as determined by SDS-PAGE. |
7265 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |