missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thiopurine S-methyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Thiopurine S-methyltransferase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8325-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifications
NP_000358.1 | |
Thiopurine S-methyltransferase Protein | |
Research Use Only | |
TPMT | |
Recombinant Protein | |
E. coli | |
TRTSLDIEEY SDTEVQKNQV LTLEEWQDKW VNGKTAFHQE QGHQLLKKHL DTFLKGKSGL RVFFPLCGKA VEMKWFADRG HSVVGVEISE LGIQEFFTEQ NLSYSEEPIT EIPGTKVFKS SSGNISLYCC SIFDLPRTNI GKFDMIWDRG ALVAINPGDR KCYADTMFSL LGKKFQYLLC VLSYDPTKHP GPPFYVPHAE IERLFGKICN IRCLEKVDAF EERHKSWGID CLFEKLYLLT E | |
Greater than 90% as determined by SDS-PAGE. |
7172 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |