Learn More
enQuireBio™ Recombinant Human TIMM21 Protein
A DNA sequence encoding the Homo sapiens (Human) TIMM21, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8108-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_054896.2 | |
TIMM21 Protein | |
Research Use Only | |
TIMM21 | |
Recombinant Protein | |
E. coli | |
MICTFLRAVQ YTEKLHRSSA KRLLLPYIVL NKACLKTEPS LRCGLQYQKK TLRPRCILGV TQKTIWTQGP SPRKAKEDGS KQVSVHRSQR GGTAVPTSQK VKEAGRDFTY LIVVLFGISI TGGLFYTIFK ELFSSSSPSK IYGRALEKCR SHPEVIGVFG ESVKGYGEVT RRGRRQHVRF TEYVKDGLKH TCVKFYIEGS EPGKQGTVYA QVKENPGSGE YDFRYIFVEI ESYPRRTIII EDNRSQDD | |
Greater than 80% as determined by SDS-PAGE. |
29090 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |