missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Toll-like receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Toll-like receptor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9436-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_003254.2 | |
Toll-like receptor 1 Protein | |
Research Use Only | |
TLR1 | |
Recombinant Protein | |
Yeast | |
SEFLVDRSKN GLIHVPKDLS QKTTILNISQ NYISELWTSD ILSLSKLRIL IISHNRIQYL DISVFKFNQE LEYLDLSHNK LVKISCHPTV NLKHLDLSFN AFDALPICKE FGNMSQLKFL GLSTTHLEKS SVLPIAHLNI SKVLLVLGET YGEKEDPEGL QDFNTESLHI VFPTNKEFHF ILDVSVKTVA NLELSNIKCV LEDNKCSYFL SILAKLQTNP KLSNLTLNNI ETTWNSFIRI LQLVWHTTVW YFSISNVKLQ GQLDFRDFDY SGTSLKALSI HQVVSDVFGF PQSYIYEIFS NMNIKNFTVS GTRMVHMLCP SKISPFLHLD FSNNLLTDTV FENCGHLTEL ETLILQMNQL KELSKIAEMT TQMKSLQQLD ISQNSVSYDE KKGDCSWTKS LLSLNMSSNI LTDTIFRCLP PRIKVLDLHS NKIKSIPKQV VKLEALQELN VAFNSLTDLP GCGSFSSLSV LIIDHNSVSH PSADFFQSCQ KMRSIKAGDN PFQCTCELGE FVKNIDQVSS EVLEGWPDSY KCDYPESYRG TLLKDFHMSE LSCNIT | |
Greater than 90% as determined by SDS-PAGE. |
7096 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |