missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TOMM20 Protein

A cDNA sequence encoding the TOMM20 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13782-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15944689

  • 181.00€ / 10µg

Spedizione stimata: 07-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

TOMM20 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TOMM20 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 20% glycerol and 2mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSDRKRRSD PNFKNRLRER RKKQKLAKER AGLSKLPDLK DAEAVQKFFL EEIQLGEELL AQGEYEKGVD HLTNAIAVCG QPQQLLQVLQ QTLPPPVFQM LLTKLPTISQ RIVSAQSLAE DDVE
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali