missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TPSAB1 Protein
A cDNA sequence encoding the TPSAB1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13798-HIS-INSECT-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
TPSAB1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
IVGGQEAPRS KWPWQVSLRV HGPYWMHFCG GSLIHPQWVL TAAHCVGPDV KDLAALRVQL REQHLYYQDQ LLPVSRIIVH PQFYTAQIGA DIALLELEEP VNVSSHVHTV TLPPASETFP PGMPCWVTGW GDVDNDERLP PPFPLKQVKV PIMENHICDA KYHLGAYTGD DVRIVRDDML CAGNTRRDSC QGDSGGPLVC KVNGTWLQAG VVSWGEGCAQ PNRPGIYTRV TYYLDWIHHY VPKKPHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TPSAB1 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 20% glycerol. |