missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRPT1 Protein
A cDNA sequence encoding the TRPT1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13822-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
TRPT1 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TRPT1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMNFSGG GRQEAAGSRG RRAPRPREQD RDVQLSKALS YALRHGALKL GLPMGADGFV PLGTLLQLPQ FRGFSAEDVQ RVVDTNRKQR FALQLGDPST GLLIRANQGH SLQVPKLELM PLETPQALPP MLVHGTFWKH WPSILLKGLS CQGRTHIHLA PGLPGDPGII SGMRSHCEIA VFIDGPLALA DGIPFFRSAN GVILTPGNTD GFLLPKYFKE ALQLRPTRKP LSLAGDEETE CQSSPKHSSR ERRRIQQ |