missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2W Protein
A DNA sequence encoding the Homo sapiens (Human) UBE2W, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7913-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001001481.2 | |
UBE2W Protein | |
Research Use Only | |
UBE2W | |
Recombinant Protein | |
E. coli | |
MASMQKRLQK ELLALQNDPP PGMTLNEKSV QNSITQWIVD MEGAPGTLYE GEKFQLLFKF SSRYPFDSPQ VMFTGENIPV HPHVYSNGHI CLSILTEDWS PALSVQSVCL SIISMLSSCK EKRRPPDNSF YVRTCNKNPK KTKWWYHDDT C | |
Greater than 90% as determined by SDS-PAGE. |
55284 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |