missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Unconventional myosin-XIX Protein
A DNA sequence encoding the Homo sapiens (Human) Unconventional myosin-XIX, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7873-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001028752.1 | |
Unconventional myosin-XIX Protein | |
Research Use Only | |
MYO19 | |
Recombinant Protein | |
E. coli | |
LQQVNGHNPG SDGQAREYLR EDLQEFLGGE VLLYKLDDLT RVNPVTLETV LRCLQARYMA DTFYTNAGCT LVALNPFKPV PQLYSPELMR EYHAAPQPQK LKPHVFTVGE QTYRNVKSLI EPVNQSIVVS GESGAGKTWT SRCLMKFYAV VATSPASWES HKIAERIEQR ILNSNPVMEA FGNACTLRNN NSSRFGKFIQ LQLNRAQQMT GAAVQTYLLE KTRVACQASS ERNFHIFYQI CKGASEDERL QWHLPEGAAF SWLPNPERSL EEDCFEVTRE AMLHLGIDTP TQNNIFKVLA GLLHLGNIQF AASEDEAQPC QPMDDAKYSV RTAASLLGLP EDVLLEMVQI RTIRAGRQQQ VFRKPCARAE CDTRRDCLAK LIYARLFDWL VSVINSSICA DTDSWTTFIG LLDVYGFESF PDNSLEQLCI NYANEKLQLE ACGLVETIHI SAAGFPIRVS HRNFVERYKL LRRLHPCTSS GPDSPYPAKG LPEWCPHSEE ATLEPLIQDI LHTLPVLTQA AAITGDSAEA MPAPMHCGRT KVFMTDSMLE LLECGRARVL EQCARCIQGG WRRHRHREQE RQWRAVMLIQ AAIRSWLTRK HIQRLHAAAT VIKRAWQKWR IRMACLAAKE LDGVEEKHFS QAPCSLSTSP LQTRLLEAII RLWPLGLVLA NTAMGVGSFQ RKLVVWACLQ LPRGSPSSYT VQTAQDQAGV TSIRALPQGS IKFHCRKSPL RYADICPEPS PYSITGFNQI LLERHRLIHV TSSAFTGLG | |
Greater than 90% as determined by SDS-PAGE. |
80179 | |
50 μg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |