missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human USH1C Protein
A DNA sequence encoding the Homo sapiens (Human) USH1C, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8285-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001284693.1 | |
USH1C Protein | |
Research Use Only | |
USH1C | |
Recombinant Protein | |
E. coli | |
MDRKVAREFR HKVDFLIEND AEKDYLYDVL RMYHQTMDVA VLVGDLKLVI NEPSRLPLFD AIRPLIPLKH QVEYDQLTPR RSRKLKEVRL DRLHPEGLGL SVRGGLEFGC GLFISHLIKG GQADSVGLQV GDEIVRINGY SISSCTHEEV INLIRTKKTV SIKVRHIGLI PVKSSPDEPL TWQYVDQFVS ESGGVRGSLG SPGNRENKEK KVFISLVGSR GLGCSISSGP IQKPGIFISH VKPGSLSAEV GLEIGDQIVE VNGVDFSNLD HKEGRELFMT DRERLAEARQ RELQRQELLM QKRLAMESNK ILQEQQEMER QRRKEIAQKA AEENERYRKE MEQIVEEEEK FKKQWEEDWG SKEQLLLPKT ITAEVHPVPL RKPKYDQGVE PELEPADDLD GGTEEQGEQD FRKYEEGFDP YSMFTPEQIM GKDVRLLRIK KEGSLDLALE GGVDSPIGKV VVSAVYERGA AERHGGIVKG DEIMAINGKI VTDYTLAEAE AALQKAWNQG GDWIDLVVAV CPPKEYDDEL TFF | |
Greater than 90% as determined by SDS-PAGE. |
10083 | |
50 μg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |