missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vasohibin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Vasohibin-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7784-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001129946.1 | |
Vasohibin-2 Protein | |
Research Use Only | |
VASH2 | |
Recombinant Protein | |
E. coli | |
MTGSAADTHR CPHPKGAKGT RSRSSHARPV SLATSGGSEE EDKDGGVLFH VNKSGFPIDS HTWERMWMHV AKVHPKGGEM VGAIRNAAFL AKPSIPQVPN YRLSMTIPDW LQAIQNYMKT LQYNHTGTQF FEIRKMRPLS GLMETAKEMT RESLPIKCLE AVILGIYLTN GQPSIERFPI SFKTYFSGNY FHHVVLGIYC NGRYGSLGMS RRAELMDKPL TFRTLSDLIF DFEDSYKKYL HTVKKVKIGL YVPHEPHSFQ PIEWKQLVLN VSKMLRADIR KELEKYARDM RMKILKPASA HSPTQVRSRG KSLSPRRRQA SPPRRLGRRE KSPALPEKKV ADLSTLNEVG YQIRI | |
Greater than 80% as determined by SDS-PAGE. |
79805 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |