missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VSTM2L Protein
A DNA sequence encoding the Homo sapiens (Human) VSTM2L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7916-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_542174.1 | |
VSTM2L Protein | |
Research Use Only | |
VSTM2L | |
Recombinant Protein | |
E. coli | |
TRPAGHAPWD NHVSGHALFT ETPHDMTART GEDVEMACSF RGSGSPSYSL EIQWWYVRSH RDWTDKQAWA SNQLKASQQE DAGKEATKIS VVKVVGSNIS HKLRLSRVKP TDEGTYECRV IDFSDGKARH HKVKAYLRVQ PGENSVLHLP EAPPAAPAPP PPKPGKELRK RSVDQEACSL | |
Greater than 90% as determined by SDS-PAGE. |
128434 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |