missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human X-ray repair cross-complementing protein 1
A DNA sequence encoding the Homo sapiens (Human) X-ray repair cross-complementing protein 5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6894-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_066964.1 | |
X-ray repair cross-complementing protein 1 | |
Research Use Only | |
XRCC5 | |
Recombinant Protein | |
E. coli | |
LTIGSNLSIR IAAYKSILQE RVKKTWTVVD AKTLKKEDIQ KETVYCLNDD DETEVLKEDI IQGFRYGSDI VPFSKVDEEQ MKYKSEGKCF SVLGFCKSSQ VQRRFFMGNQ VLKVFAARDD EAAAVALSSL IHALDDLDMV AIVRYAYDKR ANPQVGVAFP HIKHNYECLV YVQLPFMEDL RQYMFSSLKN SKKYAPTEAQ LNAVD | |
Greater than 90% as determined by SDS-PAGE. |
7520 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |