missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger BED domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Zinc finger BED domain-containing protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8421-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001164606.1 | |
Zinc finger BED domain-containing protein 1 | |
Research Use Only | |
ALTE | |
Recombinant Protein | |
E. coli | |
MENKSLESSQ TDLKLVAHPR AKSKVWKYFG FDTNAEGCIL QWKKIYCRIC MAQIAYSGNT SNLSYHLEKN HPEEFCEFVK SNTEQMREAF ATAFSKLKPE SSQQPGQDAL AVKAGHGYDS KKQQELTAAV LGLICEGLYP ASIVDEPTFK VLLKTADPRY ELPSRKYIST KAIPEKYGAV REVILKELAE ATWCGISTDM WRSENQNRAY | |
Greater than 90% as determined by SDS-PAGE. |
9189 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |