missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 638 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 638, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6910-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001014972.1 | |
Zinc finger protein 638 Protein | |
Research Use Only | |
ZNF638 | |
Recombinant Protein | |
E. coli | |
TFATLNTKGN EGDTVRDSIG FISSQVPEDP STLVTVDEIQ DDSSDLHLVT LDEVTEEDED SLADFNNLKE ELNFVTVDEV GEEEDGDNDL KVELAQSKND HPTDKKGNRK KRAVDTKKTK LESLSQVGPV NENVMEEDLK TMIERHLTAK TPTKRVRIGK TLPSEKAVVT EPAKGEEAFQ MSEVDEESGL KDSEPERKRK KTEDSSSGKS VASDVPEELD FLVPKAGFFC PICSLFYSGE KAMTNHCKST RHKQNTEKFM AKQRKEKEQN EAEERSSR | |
Greater than 90% as determined by SDS-PAGE. |
27332 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |