missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc transporter ZIP1 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc transporter ZIP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8912-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001258886.1 | |
Zinc transporter ZIP1 Protein | |
Research Use Only | |
SLC39A1 | |
Recombinant Protein | |
E. coli | |
MEQITLAYKE QSGPSPLEET RALLGTVNGG PQHWHDGPGV PQASGAPATP SALR | |
Greater than 90% as determined by SDS-PAGE. |
27173 | |
500 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |