missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein
A DNA sequence encoding the Influenza B virus (strain B/Singapore/222/1979) Influenza B (strain B/Singapore/222/1979) Nucleoprotein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7284-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Influenza B (strain B / Singapore / 222 / 1979) NucleoProtein | |
Research Use Only | |
NP | |
Recombinant Protein | |
Yeast | |
MSNMDIDGIN TGTIDKTPEE IISGTSGATR PIIRPATLAP PSNKRTRNPS PERATTSSEA DVGRKTQKKQ TPTEIKKSVY NMVVKLGEFY NQMMVKAGLN DDMERNLIQN AHAVERILLA ATDDKKTEFQ KKKNARDVKE GKEEIDHNKT GGTFYKMVRD DKTIYFSP | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Extracellular Domain | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |