missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL-1 alpha / IL1A / IL1F1 Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-1 alpha / IL1A / IL1F1 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6213-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001036222.1 | |
IL-1 alpha / IL1A / IL1F1 Protein | |
Research Use Only | |
IL1A | |
Recombinant Protein | |
E. coli | |
SAPFSFLSNM TYHFIRIIKH EFILNDTLNQ TIIRANDQHL TAAAIHNLDE AVKFDMGAYT SSKDDTKVPV ILRISKTQLY VSAQDEDQPV LLKEMPEINK TITGSETNFL FFWETHGTKN YFISVAHPNL FIATKHDNWV CLAKGLPSIT DFQILENQA | |
Greater than 90% as determined by SDS-PAGE. |
700193 | |
50 μg | |
Not determined. | |
Full Length | |
Macaque | |
His-SUMO | |
Tris-based buffer, 50% glycerol |