missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Acetyl-coenzyme A synthetase, cytoplasmic Protein
A DNA sequence encoding the Mus musculus (Mouse) Acetyl-coenzyme A synthetase, cytoplasmic, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8182-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_062785.2 | |
Acetyl-coenzyme A synthetase, cytoplasmic Protein | |
Research Use Only | |
Acss2 | |
Recombinant Protein | |
E. coli | |
MGLPEERRKS GSGSRAREET GAEGRVRGWS PPPEVRRSAH VPSLQRYREL HRRSVEEPRE FWGNIAKEFY WKTACPGPFL QYNFDVTKGK IFTEWMKGAT TNICYNVLDR NVHEKKLGDK VAFYWEGNEP GETTKITYRE LLVQVCQFSN VLRKQGIQKG DRVAIYMPMI LELVVAMLAC ARLGALHSIV FAGFSAESLC ERILDSSCCL LITTDAFYRG EKLVNLKELA DESLEKCREK GFPVRCCIVV KHLGRAELGM NDSPSQSPPV KRPCPDVQIC WNEGVDLWWH ELMQQAGDEC EPEWCDAEDP LFILYTSGST GKPKGVVHTI GGYMLYVATT FKYVFDFHPE DVFWCTADIG WITGHSYVTY GPLANGATSV LFEGIPTYPD EGRLWSIVDK YKVTKFYTAP TAIRMLMKFG DDPVTKHSRA SLQVLGTVGE PINPEAWLWY HRVVGSQRCP IVDTFWQTET GGHMLTPLPG ATPMKPGSAS FPFFGVAPAI LNESGEELEG EAEGYLVFKQ PWPGIMRTVY GNHTRFETTY FKKFPGYYVT GDGCRRDQDG YYWITGRIDD MLNVSGHLLS TAEVESALVE HEAVAEAAVV GHPHPVKGEC LYCFVTLCDG HTFSPTLTEE LKKQIREKIG PIATPDYIQN APGLPKTRSG KIMRRVLRKI AQNDHDLGDT STVADPSVIN HLFSHRCLTT Q | |
Greater than 90% as determined by SDS-PAGE. |
60525 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |