missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Allograft inflammatory factor 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Allograft inflammatory factor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5631-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_062340.1 | |
Allograft inflammatory factor 1 Protein | |
Research Use Only | |
Aif1 | |
Recombinant Protein | |
E. coli | |
SQSRDLQGGK AFGLLKAQQE ERLEGINKQF LDDPKYSNDE DLPSKLEAFK VKYMEFDLNG NGDIDIMSLK RMLEKLGVPK THLELKRLIR EVSSGSEETF SYSDFLRMML GKRSAILRMI LMYEEKNKEH KRPTGPPAKK AISELP | |
Greater than 90% as determined by SDS-PAGE. |
11629 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |