missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Angiopoietin-2 / ANG2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Angiopoietin-2 / ANG2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8884-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_031452.2 | |
Angiopoietin-2 / ANG2 Protein | |
Research Use Only | |
Angpt2 | |
Recombinant Protein | |
E. coli | |
YSNFRKSVDS TGRRQYQVQN GPCSYTFLLP ETDSCRSSSS PYMSNAVQRD APLDYDDSVQ RLQVLENILE NNTQWLMKLE NYIQDNMKKE MVEIQQNVVQ NQTAVMIEIG TSLLNQTAAQ TRKLTDVEAQ VLNQTTRLEL QLLQHSISTN KLEKQILDQT SEINKLQNKN SFLEQKVLDM EGKHSEQLQS MKEQKDELQV LVSKQSSVID ELEKKLVTAT VNNSLLQKQQ HDLMETVNSL LTMMSSPNSK SSVAIRKEEQ TTFRDCAEIF KSGLTTSGIY TLTFPNSTEE IKAYCDMDVG GGGWTVIQHR EDGSVDFQRT WKEYKEGFGS PLGEYWLGNE FVSQLTGQHR YVLKIQLKDW EGNEAHSLYD HFYLAGEESN YRIHLTGLTG TAGKISSISQ PGSDFSTKDS DNDKCICKCS QMLSGGWWFD ACGPSNLNGQ YYPQKQNTNK FNGIKWYYWK GSGYS | |
Greater than 90% as determined by SDS-PAGE. |
11601 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |