missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Apolipoprotein A-IV Protein
A DNA sequence encoding the Mus musculus (Mouse) Apolipoprotein A-IV, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5662-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_031494.2 | |
Apolipoprotein A-IV Protein | |
Research Use Only | |
APOA4 | |
Recombinant Protein | |
E. coli | |
EVTSDQVANV VWDYFTQLSN NAKEAVEQFQ KTDVTQQLST LFQDKLGDAS TYADGVHNKL VPFVVQLSGH LAQETERVKE EIKKELEDLR DRMMPHANKV TQTFGENMQK LQEHLKPYAV DLQDQINTQT QEMKLQLTPY IQRMQTTIKE NVDNLHTSMM PLATNLKDKF NRNMEELKGH LTPRANELKA TIDQNLEDLR RSLAPLTVGV QEKLNHQMEG LAFQMKKNAE ELQTKVSAKI DQLQKNLAPL VEDVQSKVKG NTEGLQKSLE DLNRQLEQQV EEFRRTVEPM GEMFNKALVQ QLEQFRQQLG PNSGEVESHL SFLEKSLREK VNSFMSTLEK KGSPDQPQAL PLPEQAQEQA QEQAQEQVQP KPLES | |
Greater than 90% as determined by SDS-PAGE. |
11808 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |