missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C-C chemokine receptor type 5 Protein
A DNA sequence encoding the Mus musculus (Mouse) C-C chemokine receptor type 5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9040-ye-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_034047.2 | |
C-C chemokine receptor type 5 Protein | |
Research Use Only | |
Ccr5 | |
Recombinant Protein | |
Yeast | |
QEFFGLNNCS SSNRLDQAMQ ATETLGMTHC CLNPVIYAFV GEKFRSYLSV FFRKHMVKRF CKRCSIFQQD NPDRASSVYT RSTGEHEVST GL | |
Greater than 90% as determined by SDS-PAGE. |
12774 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |