missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD16 / FCGR3 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD16 / FCGR3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7302-ma-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CD16 / FCGR3 Protein | |
Research Use Only | |
FCGR3 | |
Recombinant Protein | |
Mammal | |
ALPKAVVKLD PPWIQVLKED MVTLMCEGTH NPGNSSTQWF HNGRSIRSQV QASYTFKATV NDSGEYRCQM EQTRLSDPVD LGVISDWLLL QTPQRVFLEG ETITLRCHSW RNKLLNRISF FHNEKSVRYH HYKSNFSIPK ANHSHSGDYY CKGSLGSTQH QSKPVTITVQ DPATTSSISL VWYHT | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |