missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CMPK1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CMPK1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8136-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_079923.3 | |
CMPK1 Protein | |
Research Use Only | |
Cmpk1 | |
Recombinant Protein | |
E. coli | |
MKPLVVFVLG GPGAGKGTQC ARIVEKYGYT HLSAGELLRD ERKNPDSQYG ELIEKYIKEG KIVPVEITIS LLKREMDQTM AANAQKNKFL IDGFPRNQDN LQGWNKTMDG KADVSFVLFF DCNNEICIER CLERGKSSGR SDDNRESLEK RIQTYLESTK PIIDLYEEMG KVKKIDASKS VDEVFGEVVK IFDKEG | |
Greater than 90% as determined by SDS-PAGE. |
66588 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |