Learn More
enQuireBio™ Recombinant Mouse DEFB1 / Beta-defensin 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) DEFB1 / Beta-defensin 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5926-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_031869.1 | |
DEFB1 / Beta-defensin 1 Protein | |
Research Use Only | |
Defb1 | |
Recombinant Protein | |
E. coli | |
DQYKCLQHGG FCLRSSCPSN TKLQGTCKPD KPNCCKS | |
Greater than 90% as determined by SDS-PAGE. |
13214 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |