missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse DLL4 Protein
A cDNA sequence encoding the DLL4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11670-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
DLL4 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
GSGIFQLRLQ EFVNQRGMLA NGQSCEPGCR TFFRICLKHF QATFSEGPCT FGNVSTPVLG TNSFVVRDKN SGSGRNPLQL PFNFTWPGTF SLNIQAWHTP GDDLRPETSP GNSLISQIII QGSLAVGKIW RTDEQNDTLT RLSYSYRVIC SDNYYGESCS RLCKKRDDHF GHYECQPDGS LSCLPGWTGK YCDQPICLSG CHEQNGYCSK PDECICRPGW QGRLCNECIP HNGCRHGTCS IPWQCACDEG WGGLFCDQDL NYCTHHSPCK NGSTCSNSGP KGYTCTCLPG YTGEHCELGL SKCASNPCRN GGSCKDQENS YHCLCPPGYY GQHCEHSTLT CADSPCFNGG SCRERNQGSS YACECPPNFT GSNCEKKVDR CTSNPCANGG QCQNRGPSRT CRCRPGFTGT HCELHISDCA RSPCAHGGTC HDLENGPVCT CPAGFSGRRC EVRITHDACA SGPCFNGATC YTGLSPNNFV CNCPYGFVGS RCEFPVGLPP SFPWVAHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
DLL4 protein solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |